Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide 35% (w/w) in water stabilised for analysis
Hydrogen peroxide 35% (w/w) in water stabilised for analysis
Certificates
undefined
Hydrogen peroxide 35% (w/w) in water stabilised for analysis
Certificates

Specification Test Results

  • Appearance:
    Clear solution
  • Heavy metals:
    ≤10 ppm
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    about 0.03% stabilizers
  • Residue after evaporation:
    ≤0.025 % (105°C)
  • Titration with KMnO4:
    34.5 to 37 %
  • Chloride (Cl):
    ≤2 ppm
  • Nitrates:
    ≤0.012 %
  • Phosphates:
    ≤0.012 %
  • Arsenic (As):
    ≤3 ppm
  • Chromium (Cr):
    ≤0.2 ppm
  • Iron (Fe):
    ≤0.5 ppm

Specification Test Results

Related Information