Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide 33% unstabilised, TECHNICAL
Hydrogen peroxide 33% unstabilised, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Hydrogen peroxide 33% unstabilised, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    31 - 36 %

Specification Test Results