Hydrogen peroxide 30%, extra pure
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
EINECS:231-765-0
UN:2014
ADR:5.1,II
:34.01 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient