Hydrogen peroxide 30%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Catalog # 5155-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:HDPE poly bottle, pressure bottom
- Pk:450 mL
- Seq:2675
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:25.0 - 35.0%
- Assay (H₂O₂):25.0 - 35.0 %
- Appearance:Passes Test
- Free Acid (µeq/g):≤0.5
- Chloride (Cl):≤1 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- ACS - Sulfate (SO₄):≤5 ppm
- Trace Impurities - Aluminum (Al):≤100 ppt
- Trace Impurities - Antimony (Sb):≤1000 ppt
- Trace Impurities - Arsenic (As):≤1000 ppt
- Trace Impurities - Barium (Ba):≤50 ppt
- Trace Impurities - Beryllium (Be):≤1000 ppt
- Trace Impurities - Bismuth (Bi):≤1000 ppt
- Trace Impurities - Boron (B):≤1000 ppt
- Trace Impurities - Cadmium (Cd):≤50 ppt
- Trace Impurities - Calcium (Ca):≤100 ppt
- Trace Impurities - Chromium (Cr):≤50 ppt
- Trace Impurities - Cobalt (Co):≤50 ppt
- Trace Impurities - Copper (Cu):≤50 ppt
- Trace Impurities - Gallium (Ga):≤1000 ppt
- Trace Impurities - Iron (Fe):≤100 ppt
- Trace Impurities - Lead (Pb):≤100 ppt
- Trace Impurities - Lithium (Li):≤50 ppt
- Trace Impurities - Magnesium (Mg):≤50 ppt
- Trace Impurities - Manganese (Mn):≤50 ppt
- Trace Impurities - Mercury (Hg):≤1000 ppt
- Trace Impurities - Molybdenum (Mo):≤1000 ppt
- Trace Impurities - Nickel (Ni):≤50 ppt
- Trace Impurities - Potassium (K):≤100 ppt
- Trace Impurities - Silicon (Si):≤5000 ppt
- Trace Impurities - Silver (Ag):≤50 ppt
- Trace Impurities - Sodium (Na):≤100 ppt
- Trace Impurities - Strontium (Sr):≤50 ppt
- Trace Impurities - Tin (Sn):≤50 ppt
- Trace Impurities - Titanium (Ti):≤1000 ppt
- Trace Impurities - Vanadium (V):≤1000 ppt
- Trace Impurities - Zinc (Zn):≤50 ppt
- Trace Impurities - Zirconium (Zr):≤1000 ppt
Specification Test Results
About this item
:
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
ADR:5.1,II
:34.01 g/mol
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
:GHS03
:GHS05
:GHS06
:GHS08
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333