Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide 3%, Millipore®
Hydrogen peroxide 3%, Millipore®
Catalog # SIAL88597-100ML-F
Supplier:  Merck
CAS Number:  
Hydrogen peroxide 3%, Millipore®
Catalog # SIAL88597-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Boiling Pt:100 °C (1013 hPa)
Density:1.01 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :34.01 g/mol
Storage Temperature:Ambient
EINECS:231-765-0
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333