Nitric acid 65%, EMPLURA®, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8 (5.1),II
:M-Clarity™ quality level = MQ200
EINECS:231-714-2
:GHS03
:GHS06
:GHS05
Specification Test Results
- Assay (alkalimetric):64.3 - 66.4 %
- Identity:passes test
- Appearance of solution:passes test
- Chloride (Cl):≤ 0.0003 %
- Iodate, Bromate:passes test
- Nitrogen oxides (as N₂O₃):≤ 0.003 %
- Sulphate (SO₄):≤ 0.001 %
- Heavy metals (as Pb):≤ 0.0005 %
- As (Arsenic):≤ 0.0001 %
- Ca (Calcium):≤ 0.001 %
- Fe (Iron):≤ 0.0004 %
- NH₄ (Ammonium):≤ 0.001 %
- Residual solvents (ICH Q3C):...excluded by production process
- Evaporation residue:≤ 0.01