Nitric acid 60%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS06
:GHS05
Specification Test Results
- Assay:Min. 60.0 %
- Density (20/4):1.365 - 1.375
- Heavy metals (as Pb):Max. 0.2 ppm
- Ignition residue (SO4):Max. 10 ppm
- Cl (Chloride):Max. 0.5 ppm
- PO4 (Phosphate):Max. 2 ppm
- SO4 (Sulphate):Max. 2 ppm
- As (Arsenic):Max. 0.01 ppm
- Fe (Iron):Max. 1 ppm
- Mn (Manganese):Max. 0.4 ppm
- Pb (Lead):Max. 0.05 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC20423.291 spec_VWRC20423.360
Product specification
VWR® Solvents and Acids in Plastic Bottles