Nitric acid 60%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Boiling Pt:~120 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS06
:GHS05
Specification Test Results
- Assay:Min. 60 %
- Density (20/4):1.366 - 1.380
- Heavy metals (as Pb):Max. 2 ppm
- Ignition residue (SO4):Max. 50 ppm
- Cl (Chloride):Max. 5 ppm
- SO4 (Sulphate):Max. 10 ppm
- As (Arsenic):Max. 0.05 ppm
- Fe (Iron):Max. 2 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 200L Metal drum UN (ZPACE8010201) ZPACE8010070 (25 lit)
Packaging Information
SDS_EN_CH_VWRC20421.pdf SDS_EN_DE_VWRC20421.pdf SDS_EN_NL_VWRC20421.pdf SDS_EN_UK_VWRC20421.pdf sds_VWRC20421
SDS
spec_VWRC20421.294 spec_VWRC20421.460 spec_VWRC20421.460 spec_VWRC20421.551 spec_VWRC20421.551
Product specification
VWR Chemicals Big Packs and Bulks