Order Entry
Germany
ContactUsLinkComponent
Nitric acid 52.5%, AnalaR NORMAPUR® analytical reagent
Nitric acid 52.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Nitric acid 52.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    52.5 - 54.0 %
  • Density (20/4):
    1.325 - 1.335
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Ignition residue (SO4):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 0.5 ppm
  • PO4 (Phosphate):
    Max. 2 ppm
  • SO4 (Sulphate):
    Max. 2 ppm
  • As (Arsenic):
    Max. 0.05 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Mn (Manganese):
    Max. 0.4 ppm

Specification Test Results