ContactUsLinkComponent
Antibacterial lotion and foam soaps, Stellisept® med
Antibacterial lotion and foam soaps, Stellisept® med
Catalog # BODL975665
Supplier:  Bode
undefined
Antibacterial lotion and foam soaps, Stellisept® med
Catalog # BODL975665
Supplier:  Bode
Specifications
  • Description:
    Antiseptic lotion soap, Stellisept® med, squeeze bottle
  • Seq:
    01
  • Pk:
    500 mL
Specifications
About this item
Stellisept® med is an antiseptic wash lotion that can be used for whole-body washing, hygienic handwashing and pre operative body washing.
  • Skin-friendly pH value of approximately 5,5
  • Contains no alcohol or colourants
  • Skin nurturing properties
  • Bactericidal activity
Caution: * These articles are only available in Germany.