Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydrochloric acid
Hydrochloric acid 37% analytical reagent
Hydrochloric acid 37% analytical reagent
:
Bernd Kraft
Hydrochloric acid 37% analytical reagent
:
Bernd Kraft
:
7647-01-0
:
:
HCl
:
MFCD00011324
:
110 °C (1013 hPa)
:
–30 °C
:
1.18 g/cm³ (20 °C)
:
231-595-7
:
1789
:
8,II
:
36.46 g/mol
:
GHS05
:
GHS07
:
Ambient