Order Entry
Germany
ContactUsLinkComponent
Hydrochloric acid fuming ≥37% ISO, Reag. Ph. Eur. for the determination of mercury (max. 0.005 ppm Hg), Fluka™
Irritant
Corrosive
Hydrochloric acid fuming ≥37% ISO, Reag. Ph. Eur. for the determination of mercury (max. 0.005 ppm Hg), Fluka™
Certificates
Irritant
Corrosive
Hydrochloric acid fuming ≥37% ISO, Reag. Ph. Eur. for the determination of mercury (max. 0.005 ppm Hg), Fluka™

Specification Test Results

  • Assay:
    Min. 37.0 %
  • Residue on ignition (SO₄):
    Max. 0.0005 %
  • Non-volatile matter:
    Max. 0.005 %
  • Ammonium (NH₄):
    Max. 0.0001 %
  • Aluminium (Al):
    Max. 0.000005 %
  • Arsenic (As):
    Max. 0.000001 %
  • Barium (Ba):
    Max. 0.000002 %
  • Beryllium (Be):
    Max. 0.000002 %
  • Bismuth (Bi):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000001 %
  • Cobalt (Co):
    Max. 0.000001 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00002 %
  • Germanium (Ge):
    Max. 0.000005 %
  • Mercury (Hg):
    Max. 0.0000005 %
  • Potassium (K):
    Max. 0.00001 %
  • Lithium (Li):
    Max. 0.000001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000001 %
  • Molybdenum (Mo):
    Max. 0.000002 %
  • Sodium (Na):
    Max. 0.00005 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.000002 %
  • Strontium (Sr):
    Max. 0.000001 %
  • Titanium (Ti):
    Max. 0.00001 %
  • Thallium (Tl):
    Max. 0.000005 %
  • Vanadium (V):
    Max. 0.000001 %
  • Zinc (Zn):
    Max. 0.000005 %
  • Zirconium (Zr):
    Max. 0.00001 %
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Bromide (Br):
    Max. 0.005 %
  • Free chlorine (Cl):
    Max. 0.5 ppm
  • Phosphate (PO₄):
    Max. 0.00005 %
  • Sulfate (SO₄):
    Max. 1 ppm
  • Sulfite (SO₃):
    Max. 0.0001 %
  • Appearance of the solution:
    complying
  • APHA:
    Max. 10

Specification Test Results

Related Information

Frequently Bought Together