Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution, Sigma-Aldrich®
: Merck
:151-21-3
:
:CH₃(CH₂)₁₁OSO₃Na
:MFCD00036175
:288.38 g/mol
:1…1.1 g/cm³ (20 °C)
:Ambient
:205-788-1
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals