Hydrogen peroxide solution ≥35% (RT) stabilised, Purum p.a.
Catalog # 95299-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:34.5 - 37.0 %
- Density (D 20/4):1.131 - 1.133
- Free acid (as H₂SO₄):Max. 0.02 %
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Iron (Fe):Max. 5 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 200 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Appearance (color):complying
- Appearance (form):complying
Specification Test Results
About this item
:
Boiling Pt:~ 107 °C (1013 hPa)
Melting Pt:-33 °C
Melting Pt:–33 °C
Density:1.110 g/cm³ (20 °C)
UN:2014
ADR:5.1,II
:Stabilised with Stabilised
:34.01 g/mol
EINECS:231-765-0
:GHS07
:GHS03
:GHS05
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333