Order Entry
Germany
ContactUsLinkComponent
Hydrogen peroxide 30% unstabilised, AnalaR NORMAPUR® analytical reagent
Hydrogen peroxide 30% unstabilised, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Hydrogen peroxide 30% unstabilised, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    30.0-32.0 %
  • Acidity:
    Max. 0.0002 meq/g
  • Colouration:
    Max. 10 APHA
  • Evaporation residue:
    Max. 30 ppm
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Total N (Nitrogen):
    Max. 4 ppm
  • Cl (Chloride):
    Max. 0.5 ppm
  • PO4 (Phosphate):
    Max. 2 ppm
  • SO4 (Sulphate):
    Max. 2 ppm
  • As (Arsenic):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.1 ppm

Specification Test Results