Perdrogen® solution ≥30% (w/w) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt: Dec. (on heating)
Melting Pt:–26 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:5.1,II
:Stabilised with Stabilised
:34.01 g/mol
EINECS:231-765-0
UN:2014
:GHS07
:GHS05
:GHS03
Specification Test Results
- Assay:Min. 30 %
- Non-volatile matter:Max. 0.005 %
- Residue on ignition:Max. 0.002 %
- Free acid (as H₂SO₄):Max. 0.005 %
- Acidity:complying
- Aluminium (Al):Max. 0.00005 %
- Arsenic (As):Max. 0.000001 %
- Barium (Ba):Max. 0.000005 %
- Beryllium (Be):Max. 0.000001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00002 %
- Cadmium (Cd):Max. 0.000001 %
- Cobalt (Co):Max. 0.000001 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000001 %
- Iron (Fe):Max. 0.000005 %
- Germanium (Ge):Max. 0.000005 %
- Potassium (K):Max. 0.00001 %
- Lithium (Li):Max. 0.000001 %
- Magnesium (Mg):Max. 0.000005 %
- Manganese (Mn):Max. 0.000001 %
- Molybdenum (Mo):Max. 0.000002 %
- Sodium (Na):Max. 0.00001 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.000001 %
- Strontium (Sr):Max. 0.000001 %
- Titanium (Ti):Max. 0.00001 %
- Thallium (Tl):Max. 0.000005 %
- Vanadium (V):Max. 0.000001 %
- Zinc (Zn):Max. 0.000005 %
- Zirconium (Zr):Max. 0.00001 %
- Chloride (Cl):Max. 0.00005 %
- Phosphate (PO₄):Max. 0.0001 %
- Sulfate (SO₄):Max. 0.0001 %
- Total N:Max. 0.0002 %
- Organic stabilisators:Max. 0.05 %