ContactUsLinkComponent
Pyridine-[D₅] 99 atom % D for NMR spectroscopy
Irritant
Flammable
Pyridine-[D₅] 99 atom % D for NMR spectroscopy
Certificates
undefined
Irritant
Flammable
Pyridine-[D₅] 99 atom % D for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.1 % (Coulometric)
  • D-Enrichment:
    ≥98.5 %
  • Specific optical rotation:
    +7° to +8° (20°C, 589 nm) c=5 in water