Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Naphthalene-[D8]
Naphthalene-[D₈]
Naphthalene-[D₈]
Naphthalene-[D₈]
EHERC20905100
:
EHRENSTORFER
:
Naphthalene-[D₈]
EHERC20905100
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
:
:
136.11 g/mol
:
81…83 °C
:
78 °C
:
Ambient
:
214-552-7
:
1334
:
4.1,III
:
GHS08
:
GHS07
:
GHS09
:
1146-65-2
:
C₁₀D₈
:
MFCD00001743