n-Hexane-[D₁₄] 99+ atom % D for NMR spectroscopy
:21666-38-6
:
:D₃C(CD₂)₄CD₃
:MFCD00037562
:100.07 g/mol
:69 °C (1013 hPa)
:–94.3 °C
:0.96 g/cm³ (20 °C)
:–22 °C
:1208
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Appearance:Clear colorless Liquid
- D-Enrichment:≥99.0 %