Methanol-[D₄]
Glass bottle, 1 mL
Catalog # LBCLCD4O1
Marketsource
Supplier: LABC LABORTECHNIK
Jump to:
Product Details & Documents
Synonyms: Holzalkohol-D4, Hydroxymethan-D4, Methylalkohol deuteriert, Methylalkohol-D4, Methyl-D3 Alkohol-D, Carbinol-D4
- CAS Number:811-98-3
- Formula:D₃COD
- MDL Number:MFCD00044637
- Molecular Weight:36.01 g/mol
- Boiling Pt:65,3 °C (1013 hPa)
- Melting Pt:–99 °C
- Density:0,888 g/cm³ (20 °C)
- Flash Pt:11 °C
- Storage Temperature:Raumtemperatur
- EINECS:212-378-6
- UN:1230
- ADR:3,II
Specifications
Specifications
- Packung:Glass bottle
- VE:1 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



