


Nitromethane-[D₃] (99% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13031-32-8
:
Boiling Pt:100…102 °C (760 mmHg)
Melting Pt:-26 °C
Density:1.18 g/cm³
EINECS:235-892-2
UN:1261
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (K.F.)
- D-Enrichment:≥98.5 %