

Acetonitrile-[D₃] (99% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2206-26-0
:
Formula:D₃CCN
MDL Number:MFCD00001881
:44.03 g/mol
Boiling Pt:79…81 °C (760 mmHg)
Melting Pt:–46 °C
Density:0.844 g/cm³ (20 °C)
Flash Pt:2 °C
Storage Temperature:Ambient
EINECS:218-616-5
UN:1648
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.1 % (Coulometric)
- D-Enrichment:≥98.5 %