Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glycerine
Glycerine ≥87%, Sigma-Aldrich®
Glycerine ≥87%, Sigma-Aldrich®
Glycerine ≥87%, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Glycerine ≥87%, Sigma-Aldrich®
Supplier:
Merck
Certificates
About this item
CAS Number:
56-81-5
:
Formula:
HOCH₂CH(OH)CH₂OH
MDL Number:
MFCD00004722
:
92.09 g/mol
Merck Index:
13,04497
Storage Temperature:
Ambient
EINECS:
200-289-5
Related Information
Documentation
Certificates
Search by Lot Number
Documentation