Acetic acid 30% Reag. Ph. Eur. analytical reagent, Supelco®
Catalog # 1.59166.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Explosion Limit:4 - 19.9 %(V) acetic acid
- Density:1.039 g/cm3 (20 °C)
Specification Test Results
About this item
:
:60.05 g/mol
ADR:8,
:M-Clarity™ quality level = MQ300
EINECS:200-580-7
UN:2790
:GHS05
Storage Temperature:Ambient
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152