Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Copper

1
Catalog # PROONIST-498
Copper
Catalog # PROONIST-498
CAS Number:  7440-50-8
CAS Number:  7440-50-8
Price
1 030,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63.55 g/mol
  • Merck Index:13,02545
  • Boiling Pt:2595 °C (1013 hPa)
  • Melting Pt:1083 °C
  • Density:8.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-159-6
Specifications

Specifications

  • Pk:
    1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.