Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Copper
Copper ≥99.9999% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
Copper ≥99.9999% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
Copper ≥99.9999% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
:
7440-50-8
:
:
Cu
:
MFCD00010965
:
63.55 g/mol
:
13,02545
:
2595 °C (1013 hPa)
:
1083 °C
:
8.94 g/cm³ (20 °C)
:
Ambient
:
231-159-6
msds_ALFA042980