Order Entry
Germany
ContactUsLinkComponent
Carboys, wide neck, amber, Nalgene®
Carboys, wide neck, amber, Nalgene®
Catalog # NALG2256-7020
   Low Manufacturing Impact
undefined
Carboys, wide neck, amber, Nalgene®
Catalog # NALG2256-7020
   Low Manufacturing Impact

Specifications

  • Ext.Ø:
    241 mm
  • Thread:
    83B
  • Capacity:
    10 l
  • Height:
    390 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Neck I-Ø:
    64 mm
  • Pk:
    1

Specifications

About this item

Made of HDPE, amber.

  • Good chemical resistance
  • Good temperature resistance

Protect photosensitive chemicals, reagents, buffers and standards when mixing and storing. Amber HDPE carboys with amber polypropylene (PP) closure meet the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials.