


Iodine 99+%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Merck Index:13,05036
Boiling Pt:185 °C (760 mmHg)
Melting Pt:113 °C
Density:4,9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Dark grey Metallic spheres
- Titration Iodometric:≥99.0 %
- Insoluble matter:Pass