Order Entry
Germany
Orders LinkContactUsLinkComponent
Lead

Lead

Glass bottle for solids, 100 g
Catalog # PROONCSDC70003
Health
Irritant
Environment
Lead
Catalog # PROONCSDC70003
CAS Number:  7439-92-1
Lead

Glass bottle for solids, 100 g

CAS Number:  7439-92-1
Price
442,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327 °C
  • Density:11.3437 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-100-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications