Jump to:
Product Details & Documents
- CAS Number:7439-92-1
- Formula:Pb
- MDL Number:MFCD00134050
- Molecular Weight:207.2 g/mol
- Merck Index:12,05410
- Boiling Pt:1740 °C (1013 hPa)
- Melting Pt:327 °C
- Density:11.3437 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-100-4
- ADR:9,III
- :M-Clarity™ quality level = MQ300
- UN:3077
Documents
msds_107365
SDS
ProductPage_107365
Product specification
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:327 °C
- Molar Mass:207.2 g/mol
- Boiling Point:1740 °C (1013 hPa)
- Density:11.3 g/cm3 (20 °C)
Specifications
- Pack type:Plastic bag
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



