Examination light
Catalog # 139-0181
Supplier: WALDMANN
Specifications
- Description:Halogen lamp, HX 35 P FX, UK-plug
- Pk:1
Specifications
About this item
A spring balanced arm system allows risk-free and precise positioning. The standard adaptor allows for different fixture types. It is suitable for use in clinics, vets and doctors surgeries.
- Highly efficient and flexible
- Sturdy and functional design
- Easy to clean and to disinfect
- Smooth surface, resistant materials
- Easily replaceable lamps
IEC 60601-2-41 standard examination light.