Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ethyl acetate

Glass bottle, 2,5 L
Catalog # REGERH1013
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   REAGECON
Ethyl acetate
Catalog # REGERH1013
Supplier:  REAGECON
   Safer Human and Environmental Health
CAS Number:  141-78-6
Ethyl acetate
Ethyl acetate

Glass bottle, 2,5 L

CAS Number:  141-78-6
Price
59,70 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03792
  • Boiling Pt:77.1 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0.902 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Cat. No.:
    REGERH1013
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.