Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Viscometry Standards
Viscosity calibration standard S600
Viscosity calibration standard S600
Viscosity calibration standard S600
Catalog # PSLE2700-V13
Supplier:
POULTEN SELFE LEE
CAS Number:
Expand image
Viscosity calibration standard S600
Catalog # PSLE2700-V13
Supplier:
POULTEN SELFE LEE
CAS Number:
Specifications
Pk:
500 ml
Specifications
About this item
Product Details
Product Details