Sodium dodecyl sulphate (SDS) ≥98.0%, specially pure
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:100 °C (closed cup)
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
ADR:4.1,III
REACH:01-2119489461-32
:GHS02
:GHS07
:GHS05
Specification Test Results
- Assay (calculated on anhydrous):Min. 98.0 %
- Assay (C12 compounds)(calc.on anhydrous):Min. 98.0 %
- IR Spectrum:Passes test
- Acidity or alkalinity:Max. 0.05 meq/g
- Loss on drying (100°C):Max. 1.5 %
- Cl (Chloride):Max. 0.03 %
- PO4 (Phosphate):Max. 10 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 2 ppm
- Pb (Lead):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC442442F spec_VWRC442442F spec_VWRC442442F spec_VWRC442444H spec_VWRC442444H