2,4'-Difluorobenzophenone
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:342-25-6
:
Formula:C₁₃H₈F₂O
MDL Number:MFCD00000319
:218.2 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:22…24 °C
Density:1,24 g/cm³ (20 °C)
EINECS:206-441-7