VWR® Racks for Cryoboxes
Supplier: VWR Collection
Certificates
About this item
Cardboard
- Stable and long lasting
- Lightweight - the 20 compartment rack weighs 1,1 kg without boxes
- For cryoboxes 50 mm height
Economical, coated cardboard racks for upright freezers.