Laboratory glass cleaner, borer 11 UNIVERSAL
Supplier: Borer Chemie
Certificates
About this item
Liquid, mildly alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials in ultrasonic and soaking baths.
- Rinsable without residues and guarantees therefore consistent and error-free results
- With a broad cleaning spectrum
- Perfect results even at low concentrations
- Working solution suitable for multiple use
- Contains active chlorine as a cleaning booster and is designed to replace chromosulfuric acid
- EDTA- and NTA-free