Buffer, TAE solution 25X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 25X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1,6 L
Catalog # UPTIUP892574
Supplier: Uptima
Buffer, TAE solution 25X concentrate (TRIS-acetate-EDTA buffer)
Catalog # UPTIUP892574
Supplier: Uptima
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1,6 L
- Description:TAE buffer solution 25X concentrate (TRIS-acetate-EDTA buffer)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



