Tissue-Tek® O.C.T.™ compound
SAKU4583
:
- Pack type:Plastic bottle
- Pk:125 mL
- Description:Tissue-Tek® O.C.T.™ Compound
- Seq:02
Tissue-Tek® O.C.T.™ Compound is a watersoluble formulation consisting of glycols and resins, providing an excellent specimen matrix for cryostat sectioning at temperatures below –10 °C.
No residue is left on slides during the staining procedure, eliminating undesirable background staining.