Order Entry
Germany
ContactUsLinkComponent
Polyvinyl alcohol, M.W. 27,000 ≥98%, hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol, M.W. 27,000 ≥98%, hydrolysed, Sigma-Aldrich®
Catalog # SIAL81382-250G
Supplier:  Merck
CAS Number:  
Polyvinyl alcohol, M.W. 27,000 ≥98%, hydrolysed, Sigma-Aldrich®
Catalog # SIAL81382-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
CAS Number:9002-89-5
 :25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922