Polyvinyl alcohol, M.W. 145,000 ≥98% for synthesis, hydrolysed, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922
Density:1.3 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Specification Test Results
- Melting Point:160 - 240 °C
- Bulk Density:400 - 600 kg/m3
- Density:0.4 - 0.6 g/cm3 (20 °C)