Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pancreatin
Pancreatin
Pancreatin
Supplier:
PanReac AppliChem
Certificates
Expand image
Pancreatin
Supplier:
PanReac AppliChem
Certificates
About this item
Specifications
CAS No.:
8049-47-6
Storage conditions:
Refrigerator
Enzyme name:
Pancreatin
Specifications
Related Information
Documentation
Certificates
Search by Lot Number
Documentation