Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Sodium β-glycerophosphate pentahydrate
di-Sodium β-glycerophosphate pentahydrate
di-Sodium β-glycerophosphate pentahydrate
di-Sodium β-glycerophosphate pentahydrate
Supplier:
PanReac AppliChem
Certificates
Expand image
di-Sodium β-glycerophosphate pentahydrate
Supplier:
PanReac AppliChem
Certificates
About this item
CAS Number:
13408-09-8
:
819-83-0
Formula:
C₃H₇Na₂O₆P·5H₂O
MDL Number:
MFCD00002135
:
306.11 g/mol
EINECS:
212-464-3
Related Information
Documentation
Certificates
Search by Lot Number
Documentation