β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:606-68-8
:
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200
:709.41 g/mol
EINECS:210-123-3