X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt), Sigma-Aldrich®
Catalog # SIALB8174-5TAB
Supplier: Merck
CAS Number:
Specifications
- Pk:5 Tab.
- Seq:0905
Specifications
About this item
:
:521.79 g/mol
Melting Pt:230 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
CAS Number:114162-64-0
Formula:C₂₀H₂₆BrClN₂O₇
MDL Number:MFCD00058543