Order Entry
Germany
ContactUsLinkComponent
L(-)-Menthol
L(-)-Menthol
Catalog # EHERC14866020
Supplier:  EHRENSTORFER
CAS Number:  
L(-)-Menthol
Catalog # EHERC14866020
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :156.27 g/mol
Merck Index:13,05861
Boiling Pt:212 °C (1013 hPa)
Melting Pt:41…44 °C
Density:0.89 g/cm³ (20 °C)
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:218-690-9
 :GHS07
CAS Number:2216-51-5
Formula:C₁₀H₂₀O
MDL Number:MFCD00062979