L(-)-Menthol
Catalog # EHERC14866000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:156.27 g/mol
Merck Index:13,05861
Boiling Pt:212 °C (1013 hPa)
Melting Pt:41…44 °C
Density:0.89 g/cm³ (20 °C)
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:218-690-9
:GHS07
CAS Number:2216-51-5
Formula:C₁₀H₂₀O
MDL Number:MFCD00062979