Order Entry
Germany
Orders LinkContactUsLinkComponent
L(-)-Menthol

L(-)-Menthol

Glass bottle for solids, 100 g
Catalog # CSLO2416-100G
Irritant
Marketsource
Supplier:   CAESAR LORENTZ
L(-)-Menthol
Catalog # CSLO2416-100G
Supplier:  CAESAR LORENTZ
Marketsource
CAS Number:  2216-51-5
L(-)-Menthol

Glass bottle for solids, 100 g

CAS Number:  2216-51-5
Price
44,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol
  • CAS Number:2216-51-5
  • Formula:C₁₀H₂₀O
  • MDL Number:MFCD00062979
  • Molecular Weight:156.27 g/mol
  • Merck Index:13,05861
  • Boiling Pt:212 °C (1013 hPa)
  • Melting Pt:41…44 °C
  • Density:0.89 g/cm³ (20 °C)
  • Flash Pt:96 °C
  • Storage Temperature:Ambient
  • EINECS:218-690-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications