L(-)-Menthol
Glass bottle for solids, 500 mg
Catalog # PROOMM0388.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol
- CAS Number:2216-51-5
- Formula:C₁₀H₂₀O
- MDL Number:MFCD00062979
- Molecular Weight:156.27 g/mol
- Merck Index:13,05861
- Boiling Pt:212 °C (1013 hPa)
- Melting Pt:41…44 °C
- Density:0.89 g/cm³ (20 °C)
- Flash Pt:96 °C
- Storage Temperature:Ambient
- EINECS:218-690-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



