L(-)-Menthol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2216-51-5
:
Formula:C₁₀H₂₀O
MDL Number:MFCD00062979
:156.27 g/mol
Merck Index:13,05861
Boiling Pt:212 °C (1013 hPa)
Melting Pt:41…44 °C
Density:0.89 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:96 °C
EINECS:218-690-9
:GHS07
Storage Temperature:Ambient